missing translation for 'onlineSavingsMsg'
Learn More

Hexane, For pesticide residue analysis, Solstice

Catalog No. 6001055
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001055 1 L
6001056 2.5 L
60046345 4 x 2.5 L
6001057 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6001055 Supplier Solstice Supplier No. 344841L

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Density 0.659g/mL at 25°C
Quantity 1 L
Explosion Limit 0.077
Flash Point −26°C (−14.8°F)
Ignition Point 453°F
Linear Formula CH3(CH2)4CH3
Vapor Pressure 5.2 psi (37.7°C), 256 mmHg (37.7°C), 132 mmHg (20°C)
UN Number UN1208
Beilstein 1730733
Formula Weight 86.18g/mol
Grade CHROMASOLV
Chemical Name or Material Hexane
Show More Show Less
Hazard Statement P201-P210-P273-P301 + P310-P308 + P313-P331
RTECSNumber MN9275000