missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, CHROMASOLV™, ≥97%, LC-MS, Solstice
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities
Specifications
Specifications
| Density | 0.659g/mL at 25°C |
| Quantity | 2.5 L |
| Explosion Limit | 0.077 |
| Flash Point | −26°C (−14.8°F) |
| Ignition Point | 453°F |
| Linear Formula | CH3(CH2)4CH3 |
| Vapor Pressure | 5.2 psi (37.7°C), 132 mmHg (20°C), 256 mmHg (37.7°C) |
| UN Number | UN1208 |
| Beilstein | 1730733 |
| Formula Weight | 86.18g/mol |
| Show More |
Safety and Handling
| Hazard Statement | P201-P210-P273-P301 + P310-P308 + P313-P331 |
| RTECSNumber | MN9275000 |