missing translation for 'onlineSavingsMsg'
Learn More

Hexane, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023042
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023042 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023042 Supplier Merck Emd Millipore Supplier No. RTC00008420ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

  • Grade: Certified Reference Material, pharmaceutical secondary standard
  • Formula: C6H14
  • Formula Weight: 86.18
  • Boiling Point: 69°C
  • Melting Point: -95°C
  • Flash Point: -22°C
  • Refractive Index: 1.375 n20/D
  • Density: 0.659 g/mL
  • Storage Temperature: 2°C to 30°C
  • Traceability: USP and EP primary standards
  • Certifications: ISO 17034 and ISO/IEC 17025

Specifications

Chemical Name or Material Hexane
Boiling Point 69°C (literature)
Melting Point -95°C (literature)
Color Colorless
CAS 110-54-3
Physical Form Liquid
Quantity 20 mL
Molecular Formula C6H14
Linear Formula CH3(CH2)4 CH3
MDL Number MFCD00009520
UN Number UN1208
Beilstein 1730733
Flash Point -22°C
Synonym Hexane; n-Hexane
InChI Formula InChI=1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES CCCCCC
IUPAC Name hexane
Molecular Weight (g/mol) 86.18
Formula Weight 86.18
Grade Certified Reference Material
Product Line Supelco
Refractive Index n20/D 1.375 (literature)
Appearance Colorless Liquid
Vapor Pressure 5.2 psi (37.7°C)
Density 0.659 g/mL (at 25°C (literature)); Vapor Density: ∼3 (vs air)
Show More Show Less