missing translation for 'onlineSavingsMsg'
Learn More

Hexane, B&J Brand™, for HPLC, gradient grade, GC, pesticide residue analysis and spectrophotometry, >99.9% (>60% n-Hexane), Honeywell Burdick & Jackson

Catalog No. 6001063
Change view
Click to view available options
Quantity:
20 L
Packaging:
HDPE Nowpak™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001063 20 L HDPE Nowpak™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001063 Supplier Honeywell Chemicals Supplier No. NP21220
Only null left

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

CAS Min % >60.0000%
CAS Max % >60.00%
Density 0.659 - 0.673 g/cm3
Quantity 20 L
Flash Point −26°C
Packaging HDPE Nowpak™
UN Number UN1208
Beilstein 1730733
Boiling Point 68.7°C
Formula Weight 86.18
Percent Purity >99.9%
Grade HPLC Gradient
Chemical Name or Material Hexane, B&J Brand™, for HPLC, gradient grade, GC, pesticide residue analysis and spectrophotometry, >99.9% (>60% n-Hexane)
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hexanes