missing translation for 'onlineSavingsMsg'
Learn More

Hexane, B&J Brand™, for HPLC, gradient grade, GC, pesticide residue analysis and spectrophotometry, >99.9% (>60% n-Hexane), Honeywell Burdick & Jackson

Catalog No. p-7111761
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
Catalog No. Quantity Packaging
6001066 6 x 1 L Glass Bottle
6001065 4 x 4 L Glass Bottle
2 options

Catalog No. 6001066

Supplier: Honeywell Chemicals 2121L

Only null left

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Absorbance 0.05 AU max. at 225 nm (UV), 0.01 AU max. at 250 nm (UV), 0.005 AU max. at 275 nm (UV)
CAS Min % >60.0000%
CAS Max % >60.00%, <40.0000%
Density 0.659 - 0.673 g/cm3
Boiling Point 68.7°C
Quantity 6 x 1 L
Flash Point −26°C
Packaging Glass Bottle
UN Number UN1208
Beilstein 1730733
Formula Weight 86.18
Percent Purity >99.9%
Grade HPLC Gradient
Chemical Name or Material Hexane, B&J Brand™, for HPLC, gradient grade, GC, pesticide residue analysis and spectrophotometry, >99.9% (>60% n-Hexane)
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hexanes