missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, for HPLC, gradient grade, GC, pesticide residue analysis and spectrophotometry, >99.9% (>60% n-Hexane), Solstice
Specifications
Specifications
| CAS Min % | >60.0000% |
| CAS Max % | >60.00% |
| Density | 0.659 - 0.673 g/cm3 |
| Quantity | 20 L |
| Flash Point | −26°C |
| Packaging | Returnable NOWPAK™ II |
| UN Number | UN1208 |
| Beilstein | 1730733 |
| Boiling Point | 68.7°C |
| Formula Weight | 86.18 |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hexanes |