missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, B&J Brand™, for GC, for pesticide residue analysis, non-spectrophotometric grade, >99.9% (>60% n-Hexane), Honeywell Burdick & Jackson
$970.66 - $1068.03
Chemical Identifiers
| CAS | 110-54-3 |
|---|---|
| Molecular Formula | C6H14 |
| Molecular Weight (g/mol) | 86.18 |
| MDL Number | MFCD02179311 |
| InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
| Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
| PubChem CID | 8058 |
| ChEBI | CHEBI:29021 |
| IUPAC Name | hexane |
| SMILES | CCCCCC |
Chemical Identifiers
| 110-54-3 | |
| 86.18 | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| 8058 | |
| hexane |
| C6H14 | |
| MFCD02179311 | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| CHEBI:29021 | |
| CCCCCC |
Specifications
| 110-54-3 | |
| >60.00%,<40.0000% | |
| 68.7°C | |
| −26°C | |
| C6H14 | |
| UN1208 | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| CCCCCC | |
| 86.18 | |
| CHEBI:29021 | |
| >99.9% | |
| Hexane, B&J Brand™, for GC, for pesticide residue analysis, non-spectrophotometric grade, >99.9% (>60% n-Hexane) |
| >60.0000% | |
| 0.659 - 0.673 g/cm3 | |
| 6 x 1 L | |
| Glass Bottle | |
| MFCD02179311 | |
| 1730733 | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| hexane | |
| 8058 | |
| 86.18 | |
| Pesticide |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hexanes