missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, B&J Brand™, for GC (not for pesticide residue analysis), non-spectrophotometric grade, >99.9% (>60% n-Hexane), Honeywell Burdick & Jackson
Specifications
Specifications
| CAS Min % | >60.0000% |
| CAS Max % | >60.00%, <40.0000% |
| Density | 0.659 - 0.673 g/cm3 |
| Quantity | 200 L |
| Flash Point | −26°C |
| Packaging | Returnable Steel Drum |
| UN Number | UN1208 |
| Water | 0.01% max. |
| Beilstein | 1730733 |
| Boiling Point | 68.7°C |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hexanes |