missing translation for 'onlineSavingsMsg'
Learn More

Hexane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.9% (>60% n-Hexane), Honeywell

Catalog No. p-7112047
Change view
Click to view available options
Quantity:
19 L
4 x 4 L
204 L
Packaging:
Glass Bottle
Metal Can
Metal Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001078 4 x 4 L Glass Bottle
6001079 4 x 4 L Poly Bottle
6001076 19 L Metal Can
6001077 204 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6001078 Supplier Honeywell Inc Supplier No. LP2114
Only null left

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

CAS Min % >60.0000%
CAS Max % >60.00%, <40.0000%
Density 0.659 - 0.673 g/cm3
Quantity 4 x 4 L
Flash Point −26°C
Packaging Glass Bottle
UN Number UN1208
Beilstein 1730733
Water 0.01% max.
Solubility Information 1.4% in Water at 20°C
Boiling Point 68.7°C
Formula Weight 86.18
Percent Purity >60%
Grade ACS
Assay by GC Analysis n-Hexane
Chemical Name or Material Hexane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.9% (>60% n-Hexane)
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hexanes