Learn More
Hexane-1,6-bis(tri-n-butylammonium) dihydroxide, 20% w/w aq. soln.
CAS: 69762-88-5 | C30H68N2O2 | 488.886 g/mol
Supplier: Thermo Scientific Chemicals 041725AK
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Hexane-1,6-bis(tri-n-butylammonium) dihydroxide | |
| C30H68N2O2 | |
| 250 mL | |
| tributyl-6-tributylazaniumyl hexyl azanium,dihydroxide, n1,n1,n1,n6,n6,n6-hexabutylhexane-1,6-diaminium hydroxide, tributyl 6-tributylammonio hexyl azanium dihydroxide, n,n,n,n,n,n-hexabutylhexamethylenediammonium dihydroxide solution, hexane-1,6-bis tributylammonium dihydroxide w/w aqueous solution, n,n inverted exclamation marka-hexamethylenebis tributylammonium hydroxide, n∼1∼,n∼1∼,n∼1∼,n∼6∼,n∼6∼,n∼6∼-hexabutylhexane-1,6-bis aminium dihydroxide, n,n,n,n inverted exclamation marka,n inverted exclamation marka,n inverted exclamation marka-hexabutylhexamethylenediammonium dihydroxide solution | |
| CCCC[N+](CCCC)(CCCC)CCCCCC[N+](CCCC)(CCCC)CCCC.[OH-].[OH-] | |
| 488.886 | |
| 488.87 | |
| Liquid |
| 69762-88-5 | |
| MFCD00216633 | |
| UN3267 | |
| ALYGOOWRCDZDTJ-UHFFFAOYSA-L | |
| tributyl-[6-(tributylazaniumyl)hexyl]azanium;dihydroxide | |
| 15376281 | |
| 20% w/w aq. soln. |
Chemical Identifiers
| 69762-88-5 | |
| 488.886 | |
| ALYGOOWRCDZDTJ-UHFFFAOYSA-L | |
| 15376281 | |
| CCCC[N+](CCCC)(CCCC)CCCCCC[N+](CCCC)(CCCC)CCCC.[OH-].[OH-] |
| C30H68N2O2 | |
| MFCD00216633 | |
| tributyl-6-tributylazaniumyl hexyl azanium,dihydroxide, n1,n1,n1,n6,n6,n6-hexabutylhexane-1,6-diaminium hydroxide, tributyl 6-tributylammonio hexyl azanium dihydroxide, n,n,n,n,n,n-hexabutylhexamethylenediammonium dihydroxide solution, hexane-1,6-bis tributylammonium dihydroxide w/w aqueous solution, n,n inverted exclamation marka-hexamethylenebis tributylammonium hydroxide, n∼1∼,n∼1∼,n∼1∼,n∼6∼,n∼6∼,n∼6∼-hexabutylhexane-1,6-bis aminium dihydroxide, n,n,n,n inverted exclamation marka,n inverted exclamation marka,n inverted exclamation marka-hexabutylhexamethylenediammonium dihydroxide solution | |
| tributyl-[6-(tributylazaniumyl)hexyl]azanium;dihydroxide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H318-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only