missing translation for 'onlineSavingsMsg'
Learn More

Hexanal 98.0+%, TCI America™

Catalog No. p-7130361
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
H013325ML 25 mL
H0133100ML 100 mL
H0133500ML 500 mL
3 options

Catalog No. H013325ML

Supplier: TCI America H013325ML

Only null left

Chemical Identifiers

CAS 66-25-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00007027
InChI Key JARKCYVAAOWBJS-UHFFFAOYSA-N
Synonym caproaldehyde, hexaldehyde, caproic aldehyde, capronaldehyde, 1-hexanal, n-hexanal, hexanaldehyde, n-caproaldehyde, hexylaldehyde, aldehyde c-6
PubChem CID 6184
IUPAC Name hexanal
SMILES CCCCCC=O

Specifications

Color Yellow
Boiling Point 131°C
Quantity 25 mL
UN Number 1207
Formula Weight 100.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexanal
EINECSNumber (2)-0494
RTECSNumber MN7175000
TSCA Yes