missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanal 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Jaune |
| Boiling Point | 131°C |
| Quantity | 25 mL |
| UN Number | 1207 |
| Formula Weight | 100.16 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | Hexanal |
Sécurité et manipulation
| EINECSNumber | (2)-0494 |
| RTECSNumber | MN7175000 |
| TSCA | Yes |