missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanal Diethyl Acetal 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 90°C |
| Quantity | 25 mL |
| UN Number | 1993 |
| Formula Weight | 174.28 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Hexanal Diethyl Acetal |
Safety and Handling
| EINECSNumber | (2)-2455 |
| TSCA | Yes |