missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylphosphorous triamide, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 162481000

Catalog No. AC162481000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Hexamethylphosphorous triamide
0.8900g/mL
26°C
96% min. (GC)
C6H18N3P
[(CH3)2N]3P
MFCD00008301
0.89
Solubility in water: reacts
CN(C)P(N(C)C)N(C)C
163.21
163.2
1608-26-0
150°C
Authentic
Glass bottle
1.4650 to 1.467
100mL
01,431; 02,210; 04,247; 06,279; 09,235; 10,199
hexamethylphosphorous triamide, tris dimethylamino phosphine, hexametapil, phosphorous triamide, hexamethyl, hexamethyltriamidophosphite, hexamethyltriaminophosphine, phosphine, tris dimethylamino, bis dimethylamino phosphanyl dimethylamine, hmpt van, hexamethyl phosphorus triamide
XVDBWWRIXBMVJV-UHFFFAOYSA-N
N-[bis(dimethylamino)phosphanyl]-N-methylmethanamine
15355
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-534-4

Recommended Storage : product may develop some turbidity or precipitate

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...