Learn More
Hexamethylphosphorous triamide, 97%
CAS: 1608-26-0 | C6H18N3P | 163.21 g/mol
Supplier: Thermo Scientific Chemicals A1257130
Description
Hexamethylphosphorous triamide is used as a reagent in organic synthesis as a phosphorylating agent. It is associated with carbon tetrachloride for the substitution of hydroxy groups with chlorides. It is involved in the preparation of epoxides and arene oxides from aldehydes and aryldialdehdyes respectively. It is used in the preparation of carbonates as well as reduction of ozonides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHexamethylphosphorous triamide is used as a reagent in organic synthesis as a phosphorylating agent. It is associated with carbon tetrachloride for the substitution of hydroxy groups with chlorides. It is involved in the preparation of epoxides and arene oxides from aldehydes and aryldialdehdyes respectively. It is used in the preparation of carbonates as well as reduction of ozonides.
Solubility
Miscible with water.
Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
| Hexamethylphosphorous triamide | |
| -44°C | |
| 162°C to 164°C | |
| C6H18N3P | |
| MFCD00008301 | |
| UN1993 | |
| Air sensitive; May form a precipitate | |
| Miscible with water. | |
| CN(C)P(N(C)C)N(C)C | |
| 163.21 | |
| 163.21 |
| 1608-26-0 | |
| 0.898 | |
| 37°C (98°F) | |
| 1.465 | |
| 250 g | |
| 906778 | |
| hexamethylphosphorous triamide, tris dimethylamino phosphine, hexametapil, phosphorous triamide, hexamethyl, hexamethyltriamidophosphite, hexamethyltriaminophosphine, phosphine, tris dimethylamino, bis dimethylamino phosphanyl dimethylamine, hmpt van, hexamethyl phosphorus triamide | |
| XVDBWWRIXBMVJV-UHFFFAOYSA-N | |
| N-[bis(dimethylamino)phosphanyl]-N-methylmethanamine | |
| 15355 | |
| 97% |
Chemical Identifiers
| 1608-26-0 | |
| 163.21 | |
| XVDBWWRIXBMVJV-UHFFFAOYSA-N | |
| 15355 | |
| CN(C)P(N(C)C)N(C)C |
| C6H18N3P | |
| MFCD00008301 | |
| hexamethylphosphorous triamide, tris dimethylamino phosphine, hexametapil, phosphorous triamide, hexamethyl, hexamethyltriamidophosphite, hexamethyltriaminophosphine, phosphine, tris dimethylamino, bis dimethylamino phosphanyl dimethylamine, hmpt van, hexamethyl phosphorus triamide | |
| N-[bis(dimethylamino)phosphanyl]-N-methylmethanamine |
Safety and Handling
GHS H Statement
H340-H350-H226
May cause genetic defects.
May cause cancer.
Flammable liquid and vapour.
P201-P202-P210-P233-P240-P243-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P374-P380-P501c
H226-H315-H319-H335-H351
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 216-534-4
RTECSNumber : TH3390000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only