missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylenetetramine 99.0+%, TCI America™

Catalog No. p-7130334
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H009325G 25 g
H0093500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H009325G Supplier TCI America Supplier No. H009325G
Only null left

Chemical Identifiers

CAS 100-97-0
Molecular Formula C6H12N4
Molecular Weight (g/mol) 140.19
MDL Number MFCD00006895
InChI Key VKYKSIONXSXAKP-UHFFFAOYSA-N
Synonym methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
PubChem CID 4101
ChEBI CHEBI:6824
SMILES C1N2CN3CN1CN(C2)C3

Specifications

Melting Point 280°C
Color White
pH 7.8 to 8.8 (50g/L H2O 25°C)
Quantity 25 g
UN Number 1328
Formula Weight 140.19
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Hexamethylenetetramine
EINECSNumber (5)-1155
RTECSNumber MN4725000
TSCA Yes