missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylenetetramine, ACS, 99%, Spectrum™ Chemical

Catalog No. 18610801
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610801 45 kg Poly Bottle
18610799 2.5 kg Poly Bottle
18610800 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18610801 Supplier Spectrum Chemical Mfg Cor Supplier No. M121745KGBL

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100-97-0
Molecular Formula C6H12N4
Molecular Weight (g/mol) 140.19
InChI Key VKYKSIONXSXAKP-UHFFFAOYSA-N
IUPAC Name 1,3,5,7-tetraazatricyclo[3.3.1.13,7]decane
SMILES C1N2CN3CN1CN(C2)C3

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.02
Packaging Poly Bottle
Quantity 45 kg
Percent Purity 0.99
Grade ACS
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Methylparaben Sodium

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.