missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylenetetramine, ACS, 99%, Spectrum™ Chemical

Catalog No. 18610798
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610798 500 g Poly Bottle
18610799 2.5 kg Poly Bottle
18610800 12 kg Poly Pail
18610801 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610798 Supplier Spectrum Chemical Mfg Cor Supplier No. M1217500GM
Only null left

Chemical Identifiers

CAS 100-97-0
Molecular Formula C6H12N4
Molecular Weight (g/mol) 140.19
InChI Key VKYKSIONXSXAKP-UHFFFAOYSA-N
IUPAC Name 1,3,5,7-tetraazatricyclo[3.3.1.1³,⁷]decane
SMILES C1N2CN3CN1CN(C2)C3

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.02
Packaging Poly Bottle
Quantity 500 g
Percent Purity 0.99
Grade ACS

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.