missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyleneimine 98.0+%, TCI America™

Catalog No. H0092500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0092500ML 500 mL
H009225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0092500ML Supplier TCI America Supplier No. H0092500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-49-9
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.177
MDL Number MFCD00006934
InChI Key ZSIQJIWKELUFRJ-UHFFFAOYSA-N
Synonym hexamethyleneimine, homopiperidine, hexahydroazepine, hexahydro-1h-azepine, perhydroazepine, 1h-azepine, hexahydro, hexamethylenimine, azacycloheptane, azepan, hexamethylene imine
PubChem CID 8119
ChEBI CHEBI:32616
IUPAC Name azepane
SMILES C1CCCNCC1

Specifications

Color Yellow
Boiling Point 138°C
Quantity 500 mL
UN Number 2493
Formula Weight 99.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Hexamethyleneimine
EINECSNumber (5)-1094
RTECSNumber CM3150000
TSCA Yes