missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyleneimine, 99%

Catalog No. p-3758162
Change view
Click to view available options
Quantity:
5 mL
25 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC120590050 5 mL Glass bottle
AC120590250 25 mL Glass bottle
AC120595000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC120590050 Supplier Thermo Scientific Chemicals Supplier No. 120590050

CAS: 111-49-9 | C6H13N | 99.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-49-9
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.17
MDL Number MFCD00006934
InChI Key ZSIQJIWKELUFRJ-UHFFFAOYSA-N
Synonym hexamethyleneimine, homopiperidine, hexahydroazepine, hexahydro-1h-azepine, perhydroazepine, 1h-azepine, hexahydro, hexamethylenimine, azacycloheptane, azepan, hexamethylene imine
PubChem CID 8119
ChEBI CHEBI:32616
IUPAC Name azepane
SMILES C1CCCNCC1

Specifications

Melting Point -37.0°C
Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 138.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4650 to 1.4670
Quantity 5 mL
Beilstein 20, 94
Specific Gravity 0.88
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in strongly alkaline materials, soluble in methanol, ethanol, benzene and ether
Formula Weight 99.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Hexamethyleneimine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 203-875-9

RUO – Research Use Only