missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Hexamethylene diisocyanate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
$541.73 - $541.73
Specifications
| CAS | 822-06-0 |
|---|---|
| Density | 1.047 g/mL (at 20°C) |
| Boiling Point | 82°C to 85°C (0.1 mmHg) |
| Molecular Formula | C8H12N2O2 |
| Linear Formula | OCN(CH2)6 NCO |
Description
Hexamethylene diisocyanate is an aliphatic diisocyanate monomer typically used to produce oligomers and prepolymers that when combined with a polyol produce light-stable polyurethane.Highly reactive 1,6-hexamethylene diisocyanate (HMDI) was used to synthesize lactic acid polymers from oligomers by the addition of 2,2G-bis(2-oxazoline) (BOX) as chain extenders. Self-healing ability was rendered to polyurethane elastomer by synthesizing alkoxyamine-based diol and reacting it with tri-functional homopolymer of HMDI and polyethylene glycol (PEG). Plastic optical fiber (POF) was prepared by the bulk homopolymerization of HMDI catalyzed by Tin(II)-2 ethylhexanoate (SnOct).Specifications
| 822-06-0 | |
| 82°C to 85°C (0.1 mmHg) | |
| OCN(CH2)6 NCO | |
| MFCD00002047 | |
| 168.19 | |
| Respiratory system |
| 1.047 g/mL (at 20°C) | |
| C8H12N2O2 | |
| n20/D 1.453 | |
| 1,6-Diisocyanatohexane | |
| ≥98.0% (GC) |