missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Hexamethylene diisocyanate

Catalog No. 526501L
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
526501L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 526501L Supplier Sigma Aldrich Supplier No. 526501L
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexamethylene diisocyanate is an aliphatic diisocyanate monomer typically used to produce oligomers and prepolymers that when combined with a polyol produce light-stable polyurethane.Highly reactive 1,6-hexamethylene diisocyanate (HMDI) was used to synthesize lactic acid polymers from oligomers by the addition of 2,2G-bis(2-oxazoline) (BOX) as chain extenders. Self-healing ability was rendered to polyurethane elastomer by synthesizing alkoxyamine-based diol and reacting it with tri-functional homopolymer of HMDI and polyethylene glycol (PEG). Plastic optical fiber (POF) was prepared by the bulk homopolymerization of HMDI catalyzed by Tin(II)-2 ethylhexanoate (SnOct).

Specifications

CAS 822-06-0
Density 1.047 g/mL (at 20°C)
Boiling Point 82°C to 85°C (0.1 mmHg)
Molecular Formula C8H12N2O2
Refractive Index n20/D 1.453
Linear Formula OCN(CH2)6 NCO
Quantity 1 L
MDL Number MFCD00002047
Synonym 1,6-Diisocyanatohexane
Molecular Weight (g/mol) 168.19
Percent Purity ≥98.0% (GC)
Biological Activity Respiratory system
Show More Show Less