missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethylditin 98.0+%, TCI America™
$67.11 - $205.05
Chemical Identifiers
| CAS | 661-69-8 |
|---|---|
| Molecular Formula | C6H18Sn2 |
| Molecular Weight (g/mol) | 327.63 |
| MDL Number | MFCD00008277 |
| InChI Key | CCRMAATUKBYMPA-UHFFFAOYSA-N |
| Synonym | hexamethylditin, hexamethyldistannane, distannane, hexamethyl, pennsalt td 5032, stannane, hexamethyldi, ch3 3sn 2, dimer, bis, tin, hexamethyldi, distannane, hexamethyl-8ci,9ci |
| PubChem CID | 6327618 |
| IUPAC Name | hexamethyldistannane |
| SMILES | C[Sn](C)(C)[Sn](C)(C)C |
Chemical Identifiers
| 661-69-8 | |
| 327.63 | |
| CCRMAATUKBYMPA-UHFFFAOYSA-N | |
| 6327618 | |
| C[Sn](C)(C)[Sn](C)(C)C |
| C6H18Sn2 | |
| MFCD00008277 | |
| hexamethylditin, hexamethyldistannane, distannane, hexamethyl, pennsalt td 5032, stannane, hexamethyldi, ch3 3sn 2, dimer, bis, tin, hexamethyldi, distannane, hexamethyl-8ci,9ci | |
| hexamethyldistannane |
Specifications
| 661-69-8 | |
| 182°C | |
| MFCD00008277 | |
| 3146 | |
| CCRMAATUKBYMPA-UHFFFAOYSA-N | |
| hexamethyldistannane | |
| 6327618 | |
| ≥98.0% (GC) | |
| Hexamethylditin |
| White-Yellow | |
| C6H18Sn2 | |
| 1 g | |
| hexamethylditin, hexamethyldistannane, distannane, hexamethyl, pennsalt td 5032, stannane, hexamethyldi, ch3 3sn 2, dimer, bis, tin, hexamethyldi, distannane, hexamethyl-8ci,9ci | |
| C[Sn](C)(C)[Sn](C)(C)C | |
| 327.63 | |
| 327.63 | |
| Crystalline Lumps |
Safety and Handling
RTECSNumber : WH8280000
TSCA : No