missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylditin 98.0+%, TCI America™

Catalog No. H13121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13121G 1 g
H13125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13121G Supplier TCI America Supplier No. H13121G

Chemical Identifiers

CAS 661-69-8
Molecular Formula C6H18Sn2
Molecular Weight (g/mol) 327.63
MDL Number MFCD00008277
InChI Key CCRMAATUKBYMPA-UHFFFAOYSA-N
Synonym hexamethylditin, hexamethyldistannane, distannane, hexamethyl, pennsalt td 5032, stannane, hexamethyldi, ch3 3sn 2, dimer, bis, tin, hexamethyldi, distannane, hexamethyl-8ci,9ci
PubChem CID 6327618
IUPAC Name hexamethyldistannane
SMILES C[Sn](C)(C)[Sn](C)(C)C

Specifications

Color White-Yellow
Boiling Point 182°C
Quantity 1 g
UN Number 3146
Formula Weight 327.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Hexamethylditin
RTECSNumber WH8280000
TSCA No