Learn More
Hexamethylditin, 99%
CAS: 661-69-8 | C6H18Sn2 | 327.63 g/mol
$89.97 - $309.80
Chemical Identifiers
| CAS | 661-69-8 |
|---|---|
| Molecular Formula | C6H18Sn2 |
| Molecular Weight (g/mol) | 327.63 |
| MDL Number | MFCD00008277 |
| InChI Key | CCRMAATUKBYMPA-UHFFFAOYSA-N |
| Synonym | hexamethylditin, hexamethyldistannane, distannane, hexamethyl, pennsalt td 5032, stannane, hexamethyldi, ch3 3sn 2, dimer, bis, tin, hexamethyldi, distannane, hexamethyl-8ci,9ci |
| PubChem CID | 6327618 |
| SMILES | C[Sn](C)(C)[Sn](C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC295850010
|
Thermo Scientific Chemicals
295850010 |
1 g | Glass bottle |
Each for $89.97
|
|
||||
|
AC295850050
|
Thermo Scientific Chemicals
295850050 |
5 g | Glass bottle |
Each for $309.80
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 661-69-8 | |
| 327.63 | |
| CCRMAATUKBYMPA-UHFFFAOYSA-N | |
| 6327618 |
| C6H18Sn2 | |
| MFCD00008277 | |
| hexamethylditin, hexamethyldistannane, distannane, hexamethyl, pennsalt td 5032, stannane, hexamethyldi, ch3 3sn 2, dimer, bis, tin, hexamethyldi, distannane, hexamethyl-8ci,9ci | |
| C[Sn](C)(C)[Sn](C)(C)C |
Specifications
| 661-69-8 | |
| Brown | |
| 62°C | |
| 98.5% min. (GC) | |
| C6H18Sn2 | |
| (CH3)3SnSn(CH3)3 | |
| 1 g | |
| 17,143 | |
| CCRMAATUKBYMPA-UHFFFAOYSA-N | |
| trimethyltin | |
| 6327618 | |
| 99% | |
| Hexamethylditin, 99% |
| 23.0°C to 24.0°C | |
| 182.0°C (756.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.5370 to 1.5410 | |
| MFCD00008277 | |
| 04, IV, 4346 | |
| hexamethylditin, hexamethyldistannane, distannane, hexamethyl, pennsalt td 5032, stannane, hexamethyldi, ch3 3sn 2, dimer, bis, tin, hexamethyldi, distannane, hexamethyl-8ci,9ci | |
| C[Sn](C)(C)[Sn](C)(C)C | |
| 327.63 | |
| 327.63 | |
| Powder |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Fatal in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTE
GHS Signal Word: Danger
EINECSNumber : 211-549-2
RUO – Research Use Only