missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylditin, 99%

Catalog No. AC295850250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC295850250 25 g Glass bottle
AC295850010 1 g Glass bottle
AC295850050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC295850250 Supplier Thermo Scientific Chemicals Supplier No. 295850250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 661-69-8 | C6H18Sn2 | 327.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 661-69-8
Molecular Formula C6H18Sn2
Molecular Weight (g/mol) 327.63
MDL Number MFCD00008277
InChI Key CCRMAATUKBYMPA-UHFFFAOYSA-N
Synonym hexamethylditin, hexamethyldistannane, distannane, hexamethyl, pennsalt td 5032, stannane, hexamethyldi, ch3 3sn 2, dimer, bis, tin, hexamethyldi, distannane, hexamethyl-8ci,9ci
PubChem CID 6327618
SMILES C[Sn](C)(C)[Sn](C)(C)C

Specifications

Melting Point 23.0°C to 24.0°C
Color Brown
Boiling Point 182.0°C (756.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5370 to 1.5410
Linear Formula (CH3)3SnSn(CH3)3
Quantity 25 g
Beilstein 04, IV, 4346
Fieser 17,143
IUPAC Name trimethyltin
Formula Weight 327.63
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Hexamethylditin, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Fatal in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTE
EINECSNumber 211-549-2

RUO – Research Use Only