missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyldisiloxane 98.0+%, TCI America™

Catalog No. H009125ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H009125ML 25 mL
H0091100ML 100 mL
H0091500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H009125ML Supplier TCI America Supplier No. H009125ML

Chemical Identifiers

CAS 107-46-0
Molecular Formula C6H18OSi2
Molecular Weight (g/mol) 162.379
MDL Number MFCD00008265
InChI Key UQEAIHBTYFGYIE-UHFFFAOYSA-N
Synonym hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide
PubChem CID 24764
ChEBI CHEBI:78002
IUPAC Name trimethyl(trimethylsilyloxy)silane
SMILES C[Si](C)(C)O[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 100°C
Quantity 25 mL
UN Number 1993
Formula Weight 162.38
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexamethyldisiloxane
EINECSNumber (2)-2956
RTECSNumber JM9237000
TSCA Yes