missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyldisiloxane, NMR grade, 99.7%

Catalog No. AAL1697022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1697022 100 g
AAL1697014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1697022 Supplier Thermo Scientific Chemicals Supplier No. L1697022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107-46-0 | C6H18OSi2 | 162.379 g/mol

Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule.

Solubility
Slightly miscible with water.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, bases and oxygen.

Chemical Identifiers

CAS 107-46-0
Molecular Formula C6H18OSi2
Molecular Weight (g/mol) 162.379
MDL Number MFCD00008265
InChI Key UQEAIHBTYFGYIE-UHFFFAOYSA-N
Synonym hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide
PubChem CID 24764
ChEBI CHEBI:78002
IUPAC Name trimethyl(trimethylsilyloxy)silane
SMILES C[Si](C)(C)O[Si](C)(C)C

Specifications

Boiling Point 100°C to 101°C
Melting Point -67°C
Quantity 100 g
Flash Point −2°C (28°F)
Odor Characteristic
Refractive Index 1.377
Density 0.764
Linear Formula (CH3)3SiOSi(CH3)3
UN Number UN1993
Beilstein 1736258
Solubility Information Slightly miscible with water.
Formula Weight 162.38
Percent Purity 99.7%
Grade NMR
Chemical Name or Material Hexamethyldisiloxane
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P271-P280-P303+P361+P353-P304+P340-P312-P314-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 203-492-7
RTECSNumber JM9237000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only