missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyldisilazane, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187952
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mL
50 mL
Catalog No. Quantity
111019816 50 mL
111018339 250 mL
2 options

Catalog No. 111019816

Supplier: Merck Emd Millipore 5261950ML

Only null left

Chemical Identifiers

MDL Number MFCD00008259
Synonym HMDS

Specifications

Boiling Point 125°C
CAS 999-97-3
Quantity 50 mL
Linear Formula (CH3)3 SiNHSi(CH3)3
Flash Point 11°C
Formula Weight 161.39
Percent Purity ≥99.0% (GC)
Grade GC Derivatization
RTECSNumber JM9230000