missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyldisilazane, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019816
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019816 50 mL
111018339 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019816 Supplier Merck Emd Millipore Supplier No. 5261950ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00008259
Synonym HMDS

Specifications

Boiling Point 125°C
CAS 999-97-3
Quantity 50 mL
Linear Formula (CH3)3 SiNHSi(CH3)3
Flash Point 11°C
Formula Weight 161.39
Percent Purity ≥99.0% (GC)
Grade GC Derivatization
RTECSNumber JM9230000