Learn More
Hexamethyldisilazane, 98+%
CAS: 999-97-3 | C6H19NSi2 | 161.395 g/mol
Supplier: Thermo Scientific Chemicals A15139AE
Description
Hexamethyldisilazane is used as a solvent in organic synthesis and organometallic chemistry. It is often used as an adhesion promoter for photoresist in photolithography. Further, it is used for the preparation of trimethylsilyl ethers from hydroxy compounds. It is used as an alternative to critical point drying during sample preparation in electron microscopy. It is added to analyte to get silylated diagnostic products during pyrolysis in gas chromatography- mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHexamethyldisilazane is used as a solvent in organic synthesis and organometallic chemistry. It is often used as an adhesion promoter for photoresist in photolithography. Further, it is used for the preparation of trimethylsilyl ethers from hydroxy compounds. It is used as an alternative to critical point drying during sample preparation in electron microscopy. It is added to analyte to get silylated diagnostic products during pyrolysis in gas chromatography- mass spectrometry.
Solubility
Miscible with methanol, chloroform, ethyl ether and benzene. Immiscible with water. Â Â Â Â Â Â Â Â Â Â
Notes
Incompatible with strong oxidizing agents and strong acids. Â
Specifications
| Hexamethyldisilazane | |
| -78°C | |
| 126°C | |
| 98+% | |
| C6H19NSi2 | |
| (CH3)3SiNHSi(CH3)3 | |
| 100 mL | |
| 635752 | |
| 14,4689 | |
| Miscible with methanol,chloroform,ethyl ether and benzene. Immiscible with water. | |
| C[Si](C)(C)N[Si](C)(C)C | |
| 161.395 | |
| CHEBI:85068 | |
| ≥98% |
| 999-97-3 | |
| 0.774 | |
| 8°C (46°F) | |
| Odorless | |
| 1.408 | |
| MFCD00008259 | |
| UN3286 | |
| Moisture sensitive | |
| hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl | |
| FFUAGWLWBBFQJT-UHFFFAOYSA-N | |
| [dimethyl-(trimethylsilylamino)silyl]methane | |
| 13838 | |
| 161.4 | |
| Liquid |
Chemical Identifiers
| 999-97-3 | |
| 161.395 | |
| FFUAGWLWBBFQJT-UHFFFAOYSA-N | |
| 13838 | |
| [dimethyl-(trimethylsilylamino)silyl]methane |
| C6H19NSi2 | |
| MFCD00008259 | |
| hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl | |
| CHEBI:85068 | |
| C[Si](C)(C)N[Si](C)(C)C |
Safety and Handling
GHS H Statement
H225-H311-H331-H314-H318-H302
Highly flammable liquid and vapour.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302+H332-H311-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
EINECSNumber : 213-668-5
RTECSNumber : JM9230000
TSCA : Yes
storageNote1 : Moisture sensitive
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only