missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyldisilane 98.0+%, TCI America™

Catalog No. H063810ML
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H063810ML 10 mL
H0638100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H063810ML Supplier TCI America Supplier No. H063810ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1450-14-2
Molecular Formula C6H18Si2
Molecular Weight (g/mol) 146.38
MDL Number MFCD00008258
InChI Key NEXSMEBSBIABKL-UHFFFAOYSA-N
Synonym hexamethyldisilane, disilane, hexamethyl, permethyldisilane, disilane, 1,1,1,2,2,2-hexamethyl, 1,1,1,2,2,2-hexamethyldisilane, trimethyl trimethylsilyl silane, ch3 6si2, hecamethyldisilane, disilane m6, hexamethyl disilane
PubChem CID 74057
IUPAC Name hexamethyldisilane
SMILES C[Si](C)(C)[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 113°C
Quantity 10 mL
UN Number 1993
Formula Weight 146.38
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexamethyldisilane
EINECSNumber (2)-3711
RTECSNumber JM9170000
TSCA Yes