missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethyldisilane 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 113°C |
| Quantity | 100 mL |
| UN Number | 1993 |
| Formula Weight | 146.38 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Hexamethyldisilane |
Safety and Handling
| EINECSNumber | (2)-3711 |
| RTECSNumber | JM9170000 |
| TSCA | Yes |