missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethyldisilane, 98+%
Description
Hexamethyldisilane acts as a silylating reagent for allylic acetates, aryl halides and diketones. It is a source material for vapor deposition during silicon carbide growth. It is also used in electronic and semiconducter industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hexamethyldisilane acts as a silylating reagent for allylic acetates, aryl halides and diketones. It is a source material for vapor deposition during silicon carbide growth. It is also used in electronic and semiconducter industries.
Solubility
Insoluble in water. Soluble in alcohol, ether and acetone.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids and bases.
Hexamethyldisilane acts as a silylating reagent for allylic acetates, aryl halides and diketones. It is a source material for vapor deposition during silicon carbide growth. It is also used in electronic and semiconducter industries.
Solubility
Insoluble in water. Soluble in alcohol, ether and acetone.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids and bases.
Specifications
Specifications
| Boiling Point | 112°C to 113°C |
| Melting Point | 12°C to 13°C |
| Quantity | 100 g |
| Flash Point | −1°C (30°F) |
| Odor | Odorless |
| Refractive Index | 1.422 |
| Density | 0.721 |
| UN Number | UN1993 |
| Beilstein | 1633463 |
| Solubility Information | Insoluble in water. Soluble in alcohol,ether and acetone. |
| Show More |
Safety and Handling
| Hazard Category | H225 |
| Hazard Statement | GHS H Statement H225-H319-H317-H335 Highly flammable liquid and vapour. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 215-911-0 |
| RTECSNumber | JM9170000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only