missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyldisilane, 98+%

Catalog No. AAA1315514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1315514 25 g
AAA1315522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1315514 Supplier Thermo Scientific Chemicals Supplier No. A1315514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1450-14-2 | C6H18Si2 | 146.38 g/mol

Hexamethyldisilane acts as a silylating reagent for allylic acetates, aryl halides and diketones. It is a source material for vapor deposition during silicon carbide growth. It is also used in electronic and semiconducter industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyldisilane acts as a silylating reagent for allylic acetates, aryl halides and diketones. It is a source material for vapor deposition during silicon carbide growth. It is also used in electronic and semiconducter industries.

Solubility
Insoluble in water. Soluble in alcohol, ether and acetone.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids and bases.

Chemical Identifiers

CAS 1450-14-2
Molecular Formula C6H18Si2
Molecular Weight (g/mol) 146.38
MDL Number MFCD00008258
InChI Key NEXSMEBSBIABKL-UHFFFAOYSA-N
Synonym hexamethyldisilane, disilane, hexamethyl, permethyldisilane, disilane, 1,1,1,2,2,2-hexamethyl, 1,1,1,2,2,2-hexamethyldisilane, trimethyl trimethylsilyl silane, ch3 6si2, hecamethyldisilane, disilane m6, hexamethyl disilane
PubChem CID 74057
IUPAC Name trimethyl(trimethylsilyl)silane
SMILES C[Si](C)(C)[Si](C)(C)C

Specifications

Boiling Point 112°C to 113°C
Melting Point 12°C to 13°C
Quantity 25 g
Flash Point −1°C (30°F)
Odor Odorless
Refractive Index 1.422
Density 0.721
UN Number UN1993
Beilstein 1633463
Solubility Information Insoluble in water. Soluble in alcohol,ether and acetone.
Formula Weight 146.38
Percent Purity ≥98%
Chemical Name or Material Hexamethyldisilane
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225-H319-H317-H335
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 215-911-0
RTECSNumber JM9170000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only