missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylcyclotrisiloxane 98.0+%, TCI America™

Catalog No. p-7130693
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
H072525G 25 g
H0725100G 100 g
H0725500G 500 g
3 options

Catalog No. H072525G

Supplier: TCI America H072525G

Only null left

Chemical Identifiers

CAS 541-05-9
Molecular Formula C6H18O3Si3
Molecular Weight (g/mol) 222.46
MDL Number MFCD00005943
InChI Key HTDJPCNNEPUOOQ-UHFFFAOYSA-N
Synonym hexamethylcyclotrisiloxane, cyclotrisiloxane, hexamethyl, dimethylsiloxane cyclic trimer, unii-epv75l8o0r, ccris 1326, sdk 10, hexamethylcyclohexasiloxane, epv75l8o0r, cyclotrisiloxane, 2,2,4,4,6,6-hexamethyl, dsstox_cid_7185
PubChem CID 10914
IUPAC Name hexamethyl-1,3,5,2,4,6-trioxatrisilinane
SMILES C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O1

Specifications

Melting Point 65°C
Color White
Boiling Point 134°C
Quantity 25 g
UN Number 1325
Formula Weight 222.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexamethylcyclotrisiloxane
EINECSNumber (7)-0475
TSCA Yes