missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethylcyclotrisiloxane, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 63.0°C to 67.0°C |
| Color | Colorless to White |
| Boiling Point | 134.0°C |
| Flash Point | 35°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass bottle |
| Quantity | 100 g |
| Beilstein | 04,III,1884 |
| Merck Index | 09,4559 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Flammable solid. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a well-ventilated place. Keep container tightly closed. |
| EINECSNumber | 208-765-4 |
| TSCA | TSCA |
RUO – Research Use Only