missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylcyclotrisiloxane, 98%

Catalog No. AC216460250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC216460250 25 g Glass bottle
AC216461000 100 g Glass bottle
AC216465000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC216460250 Supplier Thermo Scientific Chemicals Supplier No. 216460250

CAS: 541-05-9 | C6H18O3Si3 | 222.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 541-05-9
Molecular Formula C6H18O3Si3
Molecular Weight (g/mol) 222.46
MDL Number MFCD00005943
InChI Key HTDJPCNNEPUOOQ-UHFFFAOYSA-N
Synonym hexamethylcyclotrisiloxane, cyclotrisiloxane, hexamethyl, dimethylsiloxane cyclic trimer, unii-epv75l8o0r, ccris 1326, sdk 10, hexamethylcyclohexasiloxane, epv75l8o0r, cyclotrisiloxane, 2,2,4,4,6,6-hexamethyl, dsstox_cid_7185
PubChem CID 10914
SMILES C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O1

Specifications

Melting Point 63.0°C to 67.0°C
Color Colorless to White
Boiling Point 134.0°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 04,III,1884
Merck Index 09,4559
IUPAC Name 2,2,4,4,6,6-hexamethyl-1,3,5,2,4,6-trioxatrisilinane
Formula Weight 222.47
Percent Purity 98%
Physical Form Crystalline Chunks
Chemical Name or Material Hexamethylcyclotrisiloxane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.
EINECSNumber 208-765-4
TSCA TSCA

RUO – Research Use Only