missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylacetone, 98%

Catalog No. p-7035737
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5346406 5 g
AAH5346414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5346406 Supplier Thermo Scientific Chemicals Supplier No. H5346406
Only null left

CAS: 815-24-7 | C9H18O | 142.242 g/mol

Hexamethylacetone is used to prepare di-tert.-butyladamantylcarbinol by reacting with 1-bromo-adamantane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethylacetone is used to prepare di-tert.-butyladamantylcarbinol by reacting with 1-bromo-adamantane.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 815-24-7
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00008845
InChI Key UIQGEWJEWJMQSL-UHFFFAOYSA-N
PubChem CID 13152
IUPAC Name 2,2,4,4-tetramethylpentan-3-one
SMILES CC(C)(C)C(=O)C(C)(C)C

Specifications

Density 0.824
Boiling Point 152°C to 153°C
Flash Point 34°C (93°F)
Refractive Index 1.419
Quantity 5 g
UN Number UN1224
Beilstein 1701122
Synonym hexamethylacetone, 2,2,4,4-tetramethyl-3-pentanone, di-tert-butyl ketone, 3-pentanone, 2,2,4,4-tetramethyl, 2,2,4,4-tetramethyl-pentan-3-one, 3-pentanone,2,2,4,4-tetramethyl, di-t-butyl ketone, acmc-20akrk, t-c4h9 2co, 2,4,4-tetramethyl-3-pentanone
Formula Weight 142.24
Percent Purity 98%
Chemical Name or Material Hexamethylacetone
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 212-419-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only