missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethonium Chloride Dihydrate 99.0+%, TCI America™
$213.69 - $1619.07
Chemical Identifiers
| CAS | 60-25-3 |
|---|---|
| Molecular Formula | C12H32N2O2 |
| Molecular Weight (g/mol) | 236.40 |
| MDL Number | MFCD00031563 |
| InChI Key | GYLUMIIRFKDCKI-UHFFFAOYSA-L |
| Synonym | hexamethonium chloride, hexamethonium dichloride, chloor-hexaviet, esomid chloride, hexone chloride, hiohex chloride, meton, hexon chloride, methium chloride, bistrium chloride |
| PubChem CID | 93550 |
| IUPAC Name | trimethyl[6-(trimethylazaniumyl)hexyl]azanium dihydroxide |
| SMILES | [OH-].[OH-].C[N+](C)(C)CCCCCC[N+](C)(C)C |
Chemical Identifiers
| 60-25-3 | |
| 236.40 | |
| GYLUMIIRFKDCKI-UHFFFAOYSA-L | |
| 93550 | |
| [OH-].[OH-].C[N+](C)(C)CCCCCC[N+](C)(C)C |
| C12H32N2O2 | |
| MFCD00031563 | |
| hexamethonium chloride, hexamethonium dichloride, chloor-hexaviet, esomid chloride, hexone chloride, hiohex chloride, meton, hexon chloride, methium chloride, bistrium chloride | |
| trimethyl[6-(trimethylazaniumyl)hexyl]azanium dihydroxide |
Specifications
| 60-25-3 | |
| White | |
| MFCD00031563 | |
| hexamethonium chloride, hexamethonium dichloride, chloor-hexaviet, esomid chloride, hexone chloride, hiohex chloride, meton, hexon chloride, methium chloride, bistrium chloride | |
| [OH-].[OH-].C[N+](C)(C)CCCCCC[N+](C)(C)C | |
| 236.40 | |
| 273.29 | |
| Crystalline Powder |
| 292°C | |
| C12H32N2O2 | |
| 25 g | |
| GYLUMIIRFKDCKI-UHFFFAOYSA-L | |
| trimethyl[6-(trimethylazaniumyl)hexyl]azanium dihydroxide | |
| 93550 | |
| ≥99.0% (T) | |
| Hexamethonium Chloride Dihydrate |
Safety and Handling
EINECSNumber : (2)-0180
RTECSNumber : BQ8650000
TSCA : Yes