missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethonium Chloride Dihydrate 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 292°C |
| Color | White |
| Quantity | 500 g |
| Formula Weight | 273.29 |
| Percent Purity | ≥99.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Hexamethonium Chloride Dihydrate |
Safety and Handling
| EINECSNumber | (2)-0180 |
| RTECSNumber | BQ8650000 |
| TSCA | Yes |