missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethonium Bromide 98.0+%, TCI America™
$109.98 - $1047.60
Chemical Identifiers
| CAS | 55-97-0 |
|---|---|
| Molecular Formula | C12H30Br2N2 |
| Molecular Weight (g/mol) | 362.194 |
| MDL Number | MFCD00011787 |
| InChI Key | FAPSXSAPXXJTOU-UHFFFAOYSA-L |
| Synonym | hexamethonium bromide, hexamethonium dibromide, simpatoblock, gangliostat, esametina, hexameton, vegolysen, vegolysin, bistrium bromide, hexonium dibromide |
| PubChem CID | 5938 |
| IUPAC Name | trimethyl-[6-(trimethylazaniumyl)hexyl]azanium;dibromide |
| SMILES | C[N+](C)(C)CCCCCC[N+](C)(C)C.[Br-].[Br-] |
Chemical Identifiers
| 55-97-0 | |
| 362.194 | |
| FAPSXSAPXXJTOU-UHFFFAOYSA-L | |
| 5938 | |
| C[N+](C)(C)CCCCCC[N+](C)(C)C.[Br-].[Br-] |
| C12H30Br2N2 | |
| MFCD00011787 | |
| hexamethonium bromide, hexamethonium dibromide, simpatoblock, gangliostat, esametina, hexameton, vegolysen, vegolysin, bistrium bromide, hexonium dibromide | |
| trimethyl-[6-(trimethylazaniumyl)hexyl]azanium;dibromide |
Specifications
| 55-97-0 | |
| White | |
| C12H30Br2N2 | |
| 25 g | |
| FAPSXSAPXXJTOU-UHFFFAOYSA-L | |
| trimethyl-[6-(trimethylazaniumyl)hexyl]azanium;dibromide | |
| 5938 | |
| ≥98.0% (N,T) | |
| Hexamethonium Bromide |
| 285°C | |
| 6.2 - 7.0 (1% Soln.) | |
| MFCD00011787 | |
| hexamethonium bromide, hexamethonium dibromide, simpatoblock, gangliostat, esametina, hexameton, vegolysen, vegolysin, bistrium bromide, hexonium dibromide | |
| C[N+](C)(C)CCCCCC[N+](C)(C)C.[Br-].[Br-] | |
| 362.194 | |
| 362.19 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-1322
RTECSNumber : BQ8575000
TSCA : Yes