missing translation for 'onlineSavingsMsg'
Learn More

Hexamethonium Bromide 98.0+%, TCI America™

Catalog No. H048125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H048125G 25 g
H0481500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H048125G Supplier TCI America Supplier No. H048125G

Chemical Identifiers

CAS 55-97-0
Molecular Formula C12H30Br2N2
Molecular Weight (g/mol) 362.194
MDL Number MFCD00011787
InChI Key FAPSXSAPXXJTOU-UHFFFAOYSA-L
Synonym hexamethonium bromide, hexamethonium dibromide, simpatoblock, gangliostat, esametina, hexameton, vegolysen, vegolysin, bistrium bromide, hexonium dibromide
PubChem CID 5938
IUPAC Name trimethyl-[6-(trimethylazaniumyl)hexyl]azanium;dibromide
SMILES C[N+](C)(C)CCCCCC[N+](C)(C)C.[Br-].[Br-]

Specifications

Melting Point 285°C
Color White
pH 6.2 - 7.0 (1% Soln.)
Quantity 25 g
Formula Weight 362.19
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Hexamethonium Bromide
EINECSNumber (9)-1322
RTECSNumber BQ8575000
TSCA Yes