missing translation for 'onlineSavingsMsg'
Learn More

Hexamethonium bromide, 98+%

Catalog No. AAJ6046622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6046622 100 g
AAJ6046614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6046622 Supplier Thermo Scientific Chemicals Supplier No. J6046622

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nicotinic acetyl choline receptor antagonist | CAS: 55-97-0 | C12H30Br2N2 | 362.194 g/mol

Nicotinic acetyl choline receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55-97-0
Molecular Formula C12H30Br2N2
Molecular Weight (g/mol) 362.194
MDL Number MFCD00011787
InChI Key FAPSXSAPXXJTOU-UHFFFAOYSA-L
Synonym hexamethonium bromide, hexamethonium dibromide, simpatoblock, gangliostat, esametina, hexameton, vegolysen, vegolysin, bistrium bromide, hexonium dibromide
PubChem CID 5938
IUPAC Name trimethyl-[6-(trimethylazaniumyl)hexyl]azanium;dibromide
SMILES C[N+](C)(C)CCCCCC[N+](C)(C)C.[Br-].[Br-]

Specifications

Melting Point 282°C to 285°C (decomposition)
Color White
Linear Formula (CH3)3N(Br)(CH2)6N(Br)(CH3)3
Quantity 100 g
Beilstein 3715749
Merck Index 14,4688
Formula Weight 362.19
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Hexamethonium bromide
EINECSNumber 200-249-7
RTECSNumber BQ8575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only