missing translation for 'onlineSavingsMsg'
Learn More

Hexaketocyclohexane octahydrate, 99%

CAS: 7255-28-9 | C6O6 | 168.06 g/mol

$106.63 - $430.74

Chemical Identifiers

CAS 7255-28-9
Molecular Formula C6O6
Molecular Weight (g/mol) 168.06
MDL Number MFCD00001652,MFCD00149074
InChI Key PKRGYJHUXHCUCN-UHFFFAOYSA-N
Synonym cyclohexanehexone, triquinoyl, cyclohexane-1,2,3,4,5,6-hexaone, cyclohexanehexaone, hexaketocyclohexane, unii-7zr8062lfd, trichinoyl, hexaoxocyclohexane, hexaoxocyclohexaneoctahydrate, cyclohexane-hexone, octahydrate
PubChem CID 68240
IUPAC Name cyclohexane-1,2,3,4,5,6-hexone
SMILES O=C1C(=O)C(=O)C(=O)C(=O)C1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0948804
View Documents
Thermo Scientific Chemicals
L0948804
2 g
Each for $106.63
Only null left
 
AAL0948809
View Documents
Thermo Scientific Chemicals
L0948809
10 g
Each for $430.74
Only null left
 
Description

Description

Hexaketocyclohexane octahydrate is used in the preparation of hexaazatriphenylenehexacarbonitrile. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaketocyclohexane octahydrate is used in the preparation of hexaazatriphenylenehexacarbonitrile. It is used as pharmaceutical intermediates.

Notes
Store at 4°C. Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

7255-28-9
168.06
PKRGYJHUXHCUCN-UHFFFAOYSA-N
68240
O=C1C(=O)C(=O)C(=O)C(=O)C1=O
C6O6
MFCD00001652,MFCD00149074
cyclohexanehexone, triquinoyl, cyclohexane-1,2,3,4,5,6-hexaone, cyclohexanehexaone, hexaketocyclohexane, unii-7zr8062lfd, trichinoyl, hexaoxocyclohexane, hexaoxocyclohexaneoctahydrate, cyclohexane-hexone, octahydrate
cyclohexane-1,2,3,4,5,6-hexone

Specifications

7255-28-9
C6O6
2 g
cyclohexanehexone, triquinoyl, cyclohexane-1,2,3,4,5,6-hexaone, cyclohexanehexaone, hexaketocyclohexane, unii-7zr8062lfd, trichinoyl, hexaoxocyclohexane, hexaoxocyclohexaneoctahydrate, cyclohexane-hexone, octahydrate
O=C1C(=O)C(=O)C(=O)C(=O)C1=O
168.06
312.20 (168.07 Anhydrous)
Hexaketocyclohexane octahydrate
90°C to 93°C
MFCD00001652,MFCD00149074
2091040
PKRGYJHUXHCUCN-UHFFFAOYSA-N
cyclohexane-1,2,3,4,5,6-hexone
68240
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-412-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only