missing translation for 'onlineSavingsMsg'
Learn More

Hexaketocyclohexane octahydrate, 99%

Catalog No. AAL0948809
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0948809 10 g
AAL0948804 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0948809 Supplier Thermo Scientific Chemicals Supplier No. L0948809
Only null left

CAS: 7255-28-9 | C6O6 | 168.06 g/mol

Hexaketocyclohexane octahydrate is used in the preparation of hexaazatriphenylenehexacarbonitrile. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaketocyclohexane octahydrate is used in the preparation of hexaazatriphenylenehexacarbonitrile. It is used as pharmaceutical intermediates.

Notes
Store at 4°C. Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 7255-28-9
Molecular Formula C6O6
Molecular Weight (g/mol) 168.06
MDL Number MFCD00001652,MFCD00149074
InChI Key PKRGYJHUXHCUCN-UHFFFAOYSA-N
Synonym cyclohexanehexone, triquinoyl, cyclohexane-1,2,3,4,5,6-hexaone, cyclohexanehexaone, hexaketocyclohexane, unii-7zr8062lfd, trichinoyl, hexaoxocyclohexane, hexaoxocyclohexaneoctahydrate, cyclohexane-hexone, octahydrate
PubChem CID 68240
IUPAC Name cyclohexane-1,2,3,4,5,6-hexone
SMILES O=C1C(=O)C(=O)C(=O)C(=O)C1=O

Specifications

Melting Point 90°C to 93°C
Quantity 10 g
Beilstein 2091040
Formula Weight 312.20 (168.07 Anhydrous)
Percent Purity 99%
Chemical Name or Material Hexaketocyclohexane octahydrate
EINECSNumber 208-412-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only