Learn More
United Scientific™ Hexagonal Weight Sets
Ideal for classroom use.
Supplier:  United Scientific™ WHSETCF
 Afficher plus de versions de ce produit
Numéro de catalogue. S15882
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Set of 12 hexagonal steel masses with a durable black oxidized finish.
  • WHSETC-F includes eight 1oz. masses, two 4 oz. masses; two 8 oz. masses; and one 1lb. mass
  • WHSETM-F includes two each of 5 g, 10 g, 20 g, 50 g, and 200 g masses, plus one each of 100 g and 500 g masses
  • Sets come supplied in a sturdy cardboard box
Spécifications
Spécifications
Weight Set
Steel
Assorted
Customary System Weights
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS