Learn More
United Scientific™ Hexagonal Mass Set Of 5, Plated
Ideal for classroom use.
Supplier:  United Scientific™ WHXP05
Catalog No. S15935
Only null left
Explore available promotions
Description
Description
  • Set of five zinc-plated hexagonal masses supplied in a sturdy cardboard box.
  • Includes one each of 100 g, 500 g, and 1000 g, and two each of 200 g masses
Specifications
Specifications
Mass Set
Zinc
Assorted
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Warning: Cancer and reproductive harm