Learn More
Hexafluorosilicic acid, 35% w/w aq. soln.
CAS: 16961-83-4 | F6H2Si | 144.091 g/mol
Supplier: Thermo Scientific Chemicals L148290E
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
ApplicationsHexafluorosilicic acid is commonly used as a source of fluoride. It is converted to a variety of useful hexafluorosilicate salts. It is also used as an electrolyte in the Betts electrolytic process for refining lead. It is an important organic reagent for cleaving Si-O bonds of silyl ethers. Further, it is used as wood a preservation agent and also used in surface modification of calcium carbonate.
Solubility
Miscible with water.
Notes
Incompatible in strong oxidizing agents, metals, alkalis, strong acids, stoneware and glass.
Specifications
| 108°C to 109°C | |
| 16961-83-4 | |
| F6H2Si | |
| MFCD00036289 | |
| 14,4182 | |
| Miscible with water. | |
| [H+].[H+].F[Si-2](F)(F)(F)(F)F | |
| 144.091 | |
| 144.09 | |
| 1.32 |
| Hexafluorosilicic acid | |
| 2500 g | |
| H2SiF6 | |
| UN1778 | |
| hexafluorosilicic acid, fluorosilicic acid, dihydrogen hexafluorosilicate, silicate 2-, hexafluoro-, dihydrogen, hexafluorosilicon 2-; hydron, dihydrogen hexafluorosilanediuide, silicofluoridd, hexafluorosilic acid, hydrofluorosilic acid, kieselfluorwasserstoffsaure | |
| OHORFAFFMDIQRR-UHFFFAOYSA-P | |
| hexafluorosilicon(2-);hydron | |
| 21863527 | |
| 35% w/w aq. soln. |
Chemical Identifiers
| 16961-83-4 | |
| 144.091 | |
| OHORFAFFMDIQRR-UHFFFAOYSA-P | |
| 21863527 | |
| [H+].[H+].F[Si-2](F)(F)(F)(F)F |
| F6H2Si | |
| MFCD00036289 | |
| hexafluorosilicic acid, fluorosilicic acid, dihydrogen hexafluorosilicate, silicate 2-, hexafluoro-, dihydrogen, hexafluorosilicon 2-; hydron, dihydrogen hexafluorosilanediuide, silicofluoridd, hexafluorosilic acid, hydrofluorosilic acid, kieselfluorwasserstoffsaure | |
| hexafluorosilicon(2-);hydron |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: FLUOROSILICIC ACID
EINECSNumber : 241-034-8
RTECSNumber : VV8225000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only