missing translation for 'onlineSavingsMsg'
Learn More

Hexafluorosilicic acid, 35% w/w aq. soln.

Catalog No. AAL1482936
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1482936 500 g
AAL148290E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1482936 Supplier Thermo Scientific Chemicals Supplier No. L1482936

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16961-83-4 | F6H2Si | 144.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Hexafluorosilicic acid is commonly used as a source of fluoride. It is converted to a variety of useful hexafluorosilicate salts. It is also used as an electrolyte in the Betts electrolytic process for refining lead. It is an important organic reagent for cleaving Si-O bonds of silyl ethers. Further, it is used as wood a preservation agent and also used in surface modification of calcium carbonate.

Solubility
Miscible with water.

Notes
Incompatible in strong oxidizing agents, metals, alkalis, strong acids, stoneware and glass.

Chemical Identifiers

CAS 16961-83-4
Molecular Formula F6H2Si
Molecular Weight (g/mol) 144.091
MDL Number MFCD00036289
InChI Key OHORFAFFMDIQRR-UHFFFAOYSA-P
Synonym hexafluorosilicic acid, fluorosilicic acid, dihydrogen hexafluorosilicate, silicate 2-, hexafluoro-, dihydrogen, hexafluorosilicon 2-; hydron, dihydrogen hexafluorosilanediuide, silicofluoridd, hexafluorosilic acid, hydrofluorosilic acid, kieselfluorwasserstoffsaure
PubChem CID 21863527
IUPAC Name hexafluorosilicon(2-);hydron
SMILES [H+].[H+].F[Si-2](F)(F)(F)(F)F

Specifications

Boiling Point 108°C to 109°C
Quantity 500 g
Linear Formula H2SiF6
UN Number UN1778
Merck Index 14,4182
Solubility Information Miscible with water.
Formula Weight 144.09
Concentration or Composition (by Analyte or Components) 35% w/w aq. soln.
Density 1.32
Chemical Name or Material Hexafluorosilicic acid
Hazard Category H314
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: FLUOROSILICIC ACID
EINECSNumber 241-034-8
RTECSNumber VV8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only